EMAIL THIS PAGE TO A FRIEND
Problemy tuberkuleza
[Treatment of hemoptysis and hemorrhage using pyrilen and temechin in patients with pulmonary tuberculosis].
PMID 2906745
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
274631
1,2,2,6,6-Pentamethylpiperidine, 97%
C10H21N