EMAIL THIS PAGE TO A FRIEND
Water science and technology : a journal of the International Association on Water Pollution Research
Black-odor water analysis and heavy metal distribution of Yitong River in Northeast China.
PMID 29068335
Abstract
The black-odor mechanism of Yitong River was analyzed based on survey and monitoring results. We found that organic matter is the main pollution source of Yitong River and its tributaries. A black-odor evaluation model that can describe the pollution of the Yitong River water system was developed, and this model has the parameters I = 0.21 [COD