EMAIL THIS PAGE TO A FRIEND
Water science and technology : a journal of the International Association on Water Pollution Research
Synthesis of carboxyl-introduced chitosan with C
PMID 29068339
Abstract
A route for the formation of carboxyl-introduced chitosan (CI-CS) with C
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
412287
Pyromellitic dianhydride, 97%
C10H2O6