PloS one
Spatiotemporal changes in the distribution of LHFPL5 in mice cochlear hair bundles during development and in the absence of PCDH15.
PMID 29069081
Abstract
Mechanosensory transduction by vertebrate hair cells depends on a protein complex at the tips of shorter stereocilia associated with mechanoelectrical transduction channels activated by tip links in the hair bundle. In mammalian hair cells, this complex includes transmembrane channel-like protein subunit 1 (TMC1), lipoma HMGIC fusion partner-like 5 protein (LHFPL5) and protocadherin 15 (PCDH15), a lower-end component of the tip link. TMC1 interacts with LHFPL5 and PCDH15 but how the complex develops to maturity, and the relationships between these proteins, remains uncertain. Here we evaluate the spatiotemporal development of LHFPL5 distributions in mouse cochlear hair bundles by immunofluorescence and immunogold transmission electron microscopy, from postnatal day 0 (P0) through P21 in wild type and PCDH15-deficient mice. At P0, hair bundles contain many short microvilli-like processes which we term unranked stereocilia, and a subset of lengthening rows, adjacent to a kinocilium. LHFPL5 is distributed throughout the bundle, including on stereocilia tips and the kinocilium. At P3, 4-to-6 rows of ranked stereocilia are evident, total LHFPL5 expression peaks, and LHFPL5 is localised to ranked stereocilia tips of all rows and to lower shaft/ankle links. By P12, the bundle has a mature pattern with 3 ranked rows but virtually no unranked stereocilia or kinocilium; LHFPL5 expression has declined and become restricted to the tips of shorter stereocilia. Throughout development from P0, expression of LHFPL5 is greater overall on apical than basal bundles, but there is, on average, an equal amount of labelling per labelled tip. In P3 mice lacking PCDH15, LHFPL5 labelling is not at the tips but is primarily on unranked stereocilia and lower lateral links. These data show that LHFPL5 is already present in the MET apparatus at P0 but requires PCDH15 at P3 to remain there. Shaft/ankle link localisation suggests it interacts with link proteins other than PCDH15.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P5282
Phalloidin, Fluorescein Isothiocyanate Labeled, peptide from Amanita phalloides
C58H63N10O14S4