EMAIL THIS PAGE TO A FRIEND
Oncotarget
MGr1-Antigen/37 kDa laminin receptor precursor promotes cellular prion protein induced multi-drug-resistance of gastric cancer.
PMID 29069734
Abstract
Cellular prion protein (PrP
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
EHU109791 MISSION® esiRNA, esiRNA human RPSA (esiRNA1)