EMAIL THIS PAGE TO A FRIEND
Oncotarget
Endoplasmic reticulum chaperone prolyl 4-hydroxylase, beta polypeptide (P4HB) promotes malignant phenotypes in glioma via MAPK signaling.
PMID 29069756
Abstract
Endoplasmic reticulum (ER) chaperone Prolyl 4-hydroxylase, beta polypeptide (P4HB) has previously been identified as a novel target for chemoresistance in glioblastoma multiforme (GBM). Yet its functional roles in glioma carcinogenesis remain elusive. In clinical analysis using human glioma specimens and Gene Expression Omnibus (GEO) profiles, we found that aberrant expression of P4HB was correlated with high-grade malignancy and an angiogenic phenotype in glioma. Furthermore, P4HB upregulation conferred malignant characteristics including proliferation, invasion, migration and angiogenesis