EMAIL THIS PAGE TO A FRIEND
Oncotarget
Modulating BAP1 expression affects ROS homeostasis, cell motility and mitochondrial function.
PMID 29069806
Abstract
The tumor suppressor BAP1 associates with ASXL1/2 to form the core Polycomb complex PR-DUB, which catalyzes the removal of mono-ubiquitin from several substrates including histone H2A. This complex also mediates the poly-deubiquitination of HCFC1, OGT and PCG1-α, preventing them from proteasomal degradation. Surprisingly, considering its role in a Polycomb complex, no transcriptional signature was consistently found among