EMAIL THIS PAGE TO A FRIEND
Oncotarget
PDZ-containing 1 acts as a suppressor of pancreatic cancer by regulating PTEN phosphorylation.
PMID 29069834
Abstract
Phosphorylation is a recently established cause of phosphatase and tensin homolog (PTEN) gene inactivation, which leads to defect tumour-suppressor function. In pancreatic cancer, this phenomenon has not been reported. Based on database and clinical sample analyses, we found that PTEN phosphorylation occurs in pancreatic ductal adenocarcinoma patient tissues and cell lines, and we aimed to find a method for dephosphorylation. PDZ-containing 1 (PDZK1), a tumour-associated protein that shares its PDZ-binding sequence with the carboxyl-terminal domain of PTEN, was significantly down-regulated in pancreatic cancer as compared to adjacent non-tumour tissues.