EMAIL THIS PAGE TO A FRIEND
Cell discovery
Binding to RNA regulates Set1 function.
PMID 29071121
Abstract
The Set1 family of histone H3 lysine 4 (H3K4) methyltransferases is highly conserved from yeast to human. Here we show that the Set1 complex (Set1C) directly binds RNA