EMAIL THIS PAGE TO A FRIEND
Journal of microbiology (Seoul, Korea)
Spirosoma flavus sp. nov., a novel bacterium from soil of Jeju Island.
PMID 29076067
Abstract
A novel, Gram-staining negative, yellow pigmented bacterial strain, designated 15J11-2
Related Materials