EMAIL THIS PAGE TO A FRIEND
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Suppression of microRNA-454 impedes the proliferation and invasion of prostate cancer cells by promoting N-myc downstream-regulated gene 2 and inhibiting WNT/β-catenin signaling.
PMID 29080452
Abstract
MicroRNA-454 (miR-454) is emerging as critical regulator in tumorigenesis; it may function as an oncogene or a tumor suppressor. However, the role of miR-454 in prostate cancer remains unknown. In this study, we aimed to investigate the function and molecular mechanisms of miR-454 in prostate cancer. We found that miR-454 was highly expressed in prostate cancer tissues and cell lines (*p<0.05), as detected by real-time quantitative polymerase chain reaction (RT-qPCR). Cell counting kit-8 assay, colony formation assay and cell invasion assay showed that the inhibition of miR-454 significantly suppressed prostate cancer cell proliferation and invasion (*p<0.05), whereas the overexpression of miR-454 markedly promoted prostate cancer cell proliferation and invasion (*p<0.05). Bioinformatics analysis showed that N-myc downstream-regulated gene 2 (NDRG2), a well-known tumor suppressor, was identified as a potential target gene of miR-454. Dual-luciferase reporter assay showed that miR-454 directly targeted the 3'-untranslated region of NDRG2. RT-qPCR and western blot showed that miR-454 overexpression significantly decreased NDRG2 expression (*p<0.05), whereas miR-454 inhibition markedly promoted NDRG2 expression (*p<0.05). Spearman's correlation analysis showed that miR-454 expression was inversely correlated with NDRG2 expression in prostate cancer tissues (r=-0.8932; p<0.0001). Moreover, miR-454 inhibition significantly suppressed the protein expression of β-catenin (*p<0.05) and blocked the activation of WNT signaling (*p<0.05). In addition, small interfering RNA mediated NDRG2 knockdown significantly reversed the antitumor effect of miR-454 inhibition on prostate cancer cell proliferation and invasion (*p<0.05). Taken together, these results reveal an oncogenic role of miR-454, which promotes prostate cancer cell proliferation and invasion by downregulation of NDRG2. These results also suggest miR-454 as a potential therapeutic target for the treatment of prostate cancer.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
EHU066241 MISSION® esiRNA, esiRNA human NDRG2 (esiRNA1)