EMAIL THIS PAGE TO A FRIEND
Parasites & vectors
Screening and identification of host proteins interacting with Theileria annulata cysteine proteinase (TaCP) by yeast-two-hybrid system.
PMID 29084576
Abstract
Theileria annulata can infect monocytes/macrophages and B lymphocytes and causes severe lymphoproliferative disease in ruminants. Meanwhile, infection by T. annulata leads to the permanent proliferation of cell population through regulating signaling pathways of host cells. Cysteine proteinases (CPs) are one kind of protein hydrolase and usually play critical roles in parasite virulence, host invasion, nutrition and host immune response. However, the biological function of T. annulata CP (TaCP) is still unclear. In this study, a yeast-two-hybrid assay was performed to screen host proteins interacting with TaCP, to provide information to help our understanding of the molecular mechanisms between T. annulata and host cells. The cDNA from purified bovine B cells was inserted into pGADT7-SfiI vector (pGADT7-SfiI-BcDNA, Prey plasmid) for constructing the yeast two-hybrid cDNA library. TaCP was cloned into the pGBKT7 vector (pGBKT7-TaCP) and was considered as bait plasmid after evaluating the expression, auto-activation and toxicity tests in the yeast strain Y2HGold. The yeast two-hybrid screening was carried out via co-transforming bait and prey plasmids into yeast strain Y2HGold. Sequences of positive preys were analyzed using BLAST, Gene Ontology, UniProt and STRING. Two host proteins, CRBN (Bos taurus cereblon transcript variant X2) and Ppp4C (Bos indicus protein phosphatase 4 catalytic subunit) were identified to interact with TaCP. The results of functional analysis showed that the two proteins were involved in many cellular processes, such as ubiquitylation regulation, microtubule organization, DNA repair, cell apoptosis and maturation of spliceosomal snRNPs. This study is the first to screen the host proteins of bovine B cells interacting with TaCP, and 2 proteins, CRBN and Ppp4C, were identified using yeast two-hybrid technique. The results of functional analysis suggest that the two proteins are involved in many cellular processes, such as ubiquitylation regulating, microtubule organization, DNA repair, cell apoptosis and maturation of spliceosomal snRNPs. The interaction with CRBN and Ppp4C indicate that TaCP possibly is involved in regulating signaling pathways and cell proliferation, which is helpful for understanding the interaction between T. annulata and host cells.
Related Materials