EMAIL THIS PAGE TO A FRIEND
Scientific reports
Leishmania infantum lipophosphoglycan induced-Prostaglandin E
PMID 29084985
Abstract
Lipophosphoglycan (LPG) is a key virulence factor expressed on the surfaces of Leishmania promastigotes. Although LPG is known to activate macrophages, the underlying mechanisms resulting in the production of prostaglandin E
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N194
NS-398, ≥98% (HPLC), solid
C13H18N2O5S