EMAIL THIS PAGE TO A FRIEND
African health sciences
Anti-mycobacterium tuberculosis activity of polyherbal medicines used for the treatment of tuberculosis in Eastern Cape, South Africa.
PMID 29085406
Abstract
The emergence of drug-resistant strains of To evaluate the anti-tubercular activity of polyherbal medicines used for the treatment of tuberculosis. The remedies were screened against The susceptibility testing revealed that all the remedies contain anti-tubercular activity with KWTa, KWTb, KWTc, HBfs, HBts, AL and FB exhibiting more activity at a concentration below 25 µl/ml. Furthermore, MIC values exhibited inhibitory activity with the most active remedies from KWTa, HBfs and HBts at 1.562 µg/ml. However, isoniazid showed more inhibitory activity against This study has indicated that these remedies could be potential sources of new anti-mycobacterial agents against