EMAIL THIS PAGE TO A FRIEND
Iranian journal of basic medical sciences
Antigenic and immunogenic evaluation of
PMID 29085584
Abstract
The antigenic region of the The immunogenicity studies in animal models by measuring serum antibodies (IgG1, IgG2a, and IgA) and cytokines (IFN-γ and IL5) revealed that the rFlaA induces a proper immune response in animal models. The recombinant FlaA protein is antigenic and immunogenic. Therefore, it might be used in order to design of specific diagnostic kits and recombinant vaccines against