EMAIL THIS PAGE TO A FRIEND
Oncotarget
Newcastle disease virus-induced autophagy mediates antiapoptotic signaling responses in vitro and in vivo.
PMID 29088762
Abstract
In this study, we investigated the role of autophagy and apoptosis in Newcastle disease virus (NDV)-infected chicken cells and tissues. NDV-infected and starvation-induced chick embryo fibroblasts (CEF) cells showed higher autophagosome formation than mock-infected CEF cells on transmission electron microscopy. The NDV-infected CEF cells showed enhanced conversion of microtubule-associated protein 1 light chain 3-I (LC3-I) to LC3-II and degradation of p62/SQSTM1. The diminished conversion of LC3-I to LC3-II and cleaved caspase 3 and poly (ADP-ribose) polymerase (PARP) in ultraviolet-inactivated NDV-infected cells suggested that autophagosome formation was necessary for NDV replication. Inhibition of autophagy by chloroquine (CQ) enhanced apoptosis resulting in increased cleavage of caspase 3 and PARP and AnnexinV/propidium iodide staining. Autophagy induction by rapamycin resulted in upregulation of all autophagy-related genes except Beclin 1, anti-apoptosis factors, and proinflammatory cytokines in the NDV-infected spleen and lung tissues. Subsequently, decreased apoptosis was observed in NDV-infected spleens and lungs than mock-infected organs. The pan-caspase inhibitor ZVAD-FMK promoted conversion of LC3-I to LC3-II, the degradation of p62/SQSTM1, NDV replication and cell viability by inhibiting apoptosis. Our study demonstrates that apoptosis inhibition enhances autophagy and promoted cell survival and NDV replication.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
V116 Z-VAD-FMK, solid
C22H30FN3O7