EMAIL THIS PAGE TO A FRIEND
Oncotarget
Rab27A overexpression promotes bladder cancer proliferation and chemoresistance through regulation of NF-κB signaling.
PMID 29088864
Abstract
Overexpression of Rab27A has been found in human cancers. However, the clinical significance and biological effects of Rab27A in bladder cancer tissues and cell lines have not been investigated. Here, we checked Rab27A protein in 87 cases of bladder cancer using immunohistochemistry. We found that Rab27A was overexpressed in 39 of 87 (44.8%) cancer cases. Significant association was found between Rab27 and invading depth (p=0.0083). We knocked down Rab27A in 5637 cell line and transfected Rab27A plasmid in BIU-87 cell line. Rab27A depletion inhibited cell growth rate and invasion while its overexpression induced cell growth and invasion. Rab27A also promoted cancer cell growth