EMAIL THIS PAGE TO A FRIEND
Scientific reports
Combating virulence of Gram-negative bacilli by OmpA inhibition.
PMID 29089624
Abstract
Preventing the adhesion of pathogens to host cells provides an innovative approach to tackling multidrug-resistant bacteria. In this regard, the identification of outer membrane protein A (OmpA) as a key bacterial virulence factor has been a major breakthrough. The use of virtual screening helped us to identify a cyclic hexapeptide AOA-2 that inhibits the adhesion of Acinetobacter baumannii, Pseudomonas aeruginosa and Escherichia coli to host cells and the formation of biofilm, thereby preventing the development of infection in vitro and in a murine sepsis peritoneal model. Inhibition of OmpA offers a strategy as monotherapy to address the urgent need for treatments for infections caused by Gram-negative bacilli.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
233781
Triisopropylsilane, 98%
C9H22Si