EMAIL THIS PAGE TO A FRIEND
Biological & pharmaceutical bulletin
The Truncated Isoform of the Receptor Tyrosine Kinase ALK Generated by Alternative Transcription Initiation (ALK
PMID 29093346
Abstract
Anaplastic lymphoma kinase (ALK) is a receptor-type tyrosine kinase that promotes cell growth upon stimulation with ligands such as midkine and pleiotrophin. Recently, a truncated isoform of ALK was identified in a variety of tumors. This isoform is expressed from a novel ALK transcript initiated from a de novo alternative transcription initiation (ATI) site in ALK intron 19 (referred to as ALK