EMAIL THIS PAGE TO A FRIEND
Hypertension research : official journal of the Japanese Society of Hypertension
BK
PMID 29093563
Abstract
Promoter methylation is a key mechanism in the epigenetic reprogramming of gene expression patterns. Here, we investigated the contribution of DNA methylation and the associated expression and function of large-conductance Ca
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
307823
Sodium hydrogen sulfate, technical grade
HNaO4S