EMAIL THIS PAGE TO A FRIEND
Biotechnology for biofuels
A seven-gene cluster in
PMID 29093754
Abstract
Like a number of anaerobic and cellulolytic Gram-positive bacteria, the model microorganism We characterized a putative ABC transporter devoted to cellodextrins uptake, and a cellobiose phosphorylase (CbpA) in For the first time in a Gram-positive strain, we identified a three-component system and a conjugated ABC transporter/cellobiose phosphorylase system which was shown to be essential for the growth of the model cellulolytic bacterium
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C8792
Cellopentaose, ≥80%
C30H52O26
C8286
Cellotetraose, ≥85% (HPLC)
C24H42O21
C1167
D-(+)-Cellotriose, ≥93%
C18H32O16