EMAIL THIS PAGE TO A FRIEND
Oncology letters
Exogenous DKK-3/REIC inhibits Wnt/β-catenin signaling and cell proliferation in human kidney cancer KPK1.
PMID 29098038
Abstract
The third member of the Dickkopf family (DKK-3), also known as reduced expression in immortalized cells (REIC), is a tumor suppressor present in a variety of tumor cells. Regarding the regulation of the Wnt/β-catenin signaling pathway, exogenous DKK-1 and DKK-2 are reported to inhibit Wnt signaling by binding the associated effectors. However, whether exogenous DKK-3 inhibits Wnt signaling remains unclear. A recombinant protein of human full-length DKK-3 was used to investigate the exogenous effects of the protein
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
EHU027961 MISSION® esiRNA, esiRNA human LRP6 (esiRNA1)