EMAIL THIS PAGE TO A FRIEND
Oncotarget
MLACP: machine-learning-based prediction of anticancer peptides.
PMID 29100375
Abstract
Cancer is the second leading cause of death globally, and use of therapeutic peptides to target and kill cancer cells has received considerable attention in recent years. Identification of anticancer peptides (ACPs) through wet-lab experimentation is expensive and often time consuming; therefore, development of an efficient computational method is essential to identify potential ACP candidates prior to