EMAIL THIS PAGE TO A FRIEND
Oncotarget
Carbohydrate antigen 125 and carcinoembryonic antigen in the differentiation of tuberculous peritonitis and peritonitis carcinomatosa.
PMID 29100448
Abstract
Tumor markers could increase in both tuberculous peritonitis and peritonitis carcinomatosa, confusing the differentiation of these diseases. This study aimed to better understand the extent of elevation and diagnostic efficacies of carbohydrate antigen 125 (CA 125), carcinoembryonic antigen (CEA) and combinative use of them in tuberculous peritonitis and peritonitis carcinomatosa. Of 2998 patients reviewed, 101, 120 and 71 patients were assigned to TBP group (tuberculous peritonitis), non-OCA group (non-ovarian carcinoma-related peritonitis carcinomatosa) and OCA group (ovarian carcinoma-related peritonitis carcinomatosa), respectively. The composite index was calculated by CA 125 multiplying CEA. Receiver operator characteristic curves for CA 125, CEA and composite index were acquired. As a result, CA 125 value in OCA group was higher than other two groups (serum CA 125: