EMAIL THIS PAGE TO A FRIEND
Journal of pharmacological and toxicological methods
The detection and quantification, in vivo and in real time, of hydrogen sulfide in ethanol-induced lesions in rat stomachs using an ion sensitive electrode.
PMID 29100966
Abstract
The development of electrochemical sensors for the detection of small molecules has already had a significant effect on the study of biology because of their selectivity and ability to measure low concentrations of small molecules that regulate various functions in living organisms. Hydrogen sulfide (H Wistar rats fasted for 12h. Then, the control group was given an intragastrical dose of saline. l-Cysteine (50mg/kg) or dl-propargylglycine (50mg/kg) were administered to the test groups to modify the H The exogenous administration of an H This work provides the basis for a real-time analysis of the changes in-vivo of the gasotransmitter H