EMAIL THIS PAGE TO A FRIEND
Progress in neuro-psychopharmacology & biological psychiatry
Personality measures after gamma ventral capsulotomy in intractable OCD.
PMID 29100975
Abstract
Neurosurgeries such as gamma ventral capsulotomy (GVC) are an option for otherwise intractable obsessive-compulsive disorder (OCD) patients. In general, clinical and neuropsychological status both improve after GVC. However, its consequences on personality traits are not well-studied. The objective of this study was to investigate personality changes after one year of GVC in intractable OCD patients. The personality assessment was conducted using the Revised NEO Personality Inventory (NEO PI-R) and Cloninger's Temperament and Character Inventory (TCI) in 14 intractable OCD patients before and one year after GVC. Comparisons of personality features between treatment responders (n=5) and non-responders (n=9) were performed. Multiple linear regression was also used for predicting changes in clinical and global functioning variables. Overall, no deleterious effect was found in personality after GVC. Responders had a reduction in neuroticism (p=0.043) and an increase in extraversion (p=0.043). No significant changes were observed in non-responders. Increases in novelty seeking and self-directedness, and decreases in persistence and cooperativiness predicted OCD symptom improvement. Similary, improvement in functioning was also predicted by hgher novelty seeking and self-directedness after GVC, whereas better functioning was also associated with lower reward dependence and cooperativeness after surgery. The pattern of changes in personality traits after GVC was generally towards that observed in nonclinical population, and does not raise safety concerns.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
34138
Neosolaniol solution, 100 μg/mL in acetonitrile, analytical standard
C19H26O8