EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
Corrigendum to "Gas chromatographic determination of polycyclic aromatic hydrocarbons in water and smoked rice samples after solid-phase microextraction using multiwalled carbon nanotube loaded hollow fiber" [J. Chromatogr. A 1374 (2014) 50-57].
PMID 29102380