EMAIL THIS PAGE TO A FRIEND
Chinese journal of natural medicines
Neuroprotective effects of Yiqihuoxue calm wind capsule on ischemic stroke in rats.
PMID 29103461
Abstract
Stroke remains the third leading cause of death and of adult disability worldwide. Vascular occlusion, followed by ischemic cascade, leads to irreversible tissue injury. Recombinant tissue plasminogen activator is the only FDA approved drug for the current treatment of acute ischemic stroke. However, traditional Chinese medicine has a long history and rich clinical experience in the treatment and rehabilitation of ischemic stroke. Using a classical middle cerebral artery occlusion (MCAO) stroke model, we tested the effectiveness of Yiqihuoxue calm wind (YCW) capsule on neurological function, gross pathology and oxidative stress status in MCAO rats. YCW capsule (3.36 and 6.72 g·kg