EMAIL THIS PAGE TO A FRIEND
Clinical chemistry
Fructosamine: matrix problems with standards based on protein matrices.
PMID 2910573
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D6149
1-Deoxy-1-morpholino-D-fructose
C10H19NO6