EMAIL THIS PAGE TO A FRIEND
Marine environmental research
Salinity stress results in differential Hsp70 expression in the Exaiptasia pallida and Symbiodinium symbiosis.
PMID 29108677
Abstract
Abiotic factors affect cnidarian-algal symbiosis and, if severe enough, can result in bleaching. Increased temperature and light are well characterized causes of bleaching, but other factors like salinity can also stress the holobiont. In cnidarian-dinoflagellate systems, the expression of host genes, including heat shock protein 70 (Hsp70), changes due to thermal and light stress. In this experiment, we characterized to what extent salinity stress affects Hsp70 expression in the holobiont by simultaneously measuring host and symbiont Hsp70 expression in response to up to 8 h of hypo- and hypersaline conditions in the sea anemone Exaiptasia pallida and its intracellular symbiont Symbiodinium minutum. We show that E. pallida Hsp70 expression increases (6-11-fold) at high salinities whereas Symbiodinium Hsp70 expression shows little change (1.4-2.6-fold). These data suggest that cnidarian Hsp70 response is similar across multiple abiotic stressors further validating the Hsp70 gene as a biomarker for abiotic stress.