EMAIL THIS PAGE TO A FRIEND
BMC veterinary research
Ultrasonographically determined renal values and comparisons to serum biochemistry renal variables in aged semi-captive cheetahs (Acinonyx jubatus).
PMID 29110712
Abstract
Cheetahs in captivity have a high prevalence of chronic renal diseases. We ultrasonographically evaluated the renal volumes, a variety of renal dimensions, interarcuate artery resistive indices (RI) as well as aortic diameters and the length of the ventral aspect of the 6th lumbar vertebrae in 27 aged semi-captive anesthetized cheetahs. Renal size, dimensions and ratios were compared to urine specific gravity, serum creatinine and urea values. There were minimal differences for all values between left and right kidneys. Mean kidney length was 65.1 mm (range 55.2-76.9) with left kidney length ratios to L6 length being 1.60 (range 1.27-2.06) and to the aortic diameter 7.69 (range 4.54-10.72). Significant correlations between left renal length as well as length:L6 ratio to creatinine values were found ((r - 0.66) and (r - 0.60) respectively). The mean RI values of the different sedation/anesthetic protocols ranged from 0.46-0.55. Left renal length and L6 ventral vertebral body length as well as left kidney RI values should be routinely measured in all cheetah abdominal ultrasound examinations. These measurements, together with serum creatinine, urea and urine specific gravity values may be relatively sensitive indicators of early renal pathology in the absence of gross ultrasonographic changes.