EMAIL THIS PAGE TO A FRIEND
The Plant cell
EIN3 and PIF3 Form an Interdependent Module That Represses Chloroplast Development in Buried Seedlings.
PMID 29114016
Abstract
In buried seedlings, chloroplasts are arrested at the etioplast stage, but they rapidly mature upon emergence of the seedling. Etioplast-chloroplast differentiation is halted through the integration of soil-induced signals, including pressure and the absence of light, although the details on how this information converges to regulate cellular decisions remain unclear. Here, we identify an interdependent transcription module that integrates the mechanical pressure and darkness signals to control chloroplast development in