EMAIL THIS PAGE TO A FRIEND
Nature communications
Phospholipid flippase ATP11C is endocytosed and downregulated following Ca
PMID 29123098
Abstract
We and others showed that ATP11A and ATP11C, members of the P4-ATPase family, translocate phosphatidylserine (PS) and phosphatidylethanolamine from the exoplasmic to the cytoplasmic leaflets at the plasma membrane. PS exposure on the outer leaflet of the plasma membrane in activated platelets, erythrocytes, and apoptotic cells was proposed to require the inhibition of PS-flippases, as well as activation of scramblases. Although ATP11A and ATP11C are cleaved by caspases in apoptotic cells, it remains unclear how PS-flippase activity is regulated in non-apoptotic cells. Here we report that the PS-flippase ATP11C, but not ATP11A, is sequestered from the plasma membrane via clathrin-mediated endocytosis upon Ca
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G2911
GF 109203X, synthetic, ≥90% (HPLC)
C25H24N4O2