EMAIL THIS PAGE TO A FRIEND
Frontiers in molecular neuroscience
Suppression of Inflammatory Demyelinaton and Axon Degeneration through Inhibiting Kv3 Channels.
PMID 29123469
Abstract
The development of neuroprotective and repair strategies for treating progressive multiple sclerosis (MS) requires new insights into axonal injury. 4-aminopyridine (4-AP), a blocker of voltage-gated K