EMAIL THIS PAGE TO A FRIEND
The journal of physical chemistry. B
Influence of Proton Acceptors on the Proton-Coupled Electron Transfer Reaction Kinetics of a Ruthenium-Tyrosine Complex.
PMID 29130684
Abstract
A polypyridyl ruthenium complex with fluorinated bipyridine ligands and a covalently bound tyrosine moiety was synthesized, and its photo-induced proton-coupled electron transfer (PCET) reactivity in acetonitrile was investigated with transient absorption spectroscopy. Using flash-quench methodology with methyl viologen as an oxidative quencher, a Ru
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
109835
2-Methylpyridine, 98%
C6H7N