EMAIL THIS PAGE TO A FRIEND
Journal of agricultural and food chemistry
Enzymatic Synthesis and Structural Characterization of Theanderose through Transfructosylation Reaction Catalyzed by Levansucrase from Bacillus subtilis CECT 39.
PMID 29131629
Abstract
This work addresses the high-yield and fast enzymatic production of theanderose, a naturally occurring carbohydrate, also known as isomaltosucrose, whose chemical structure determined by NMR is α-d-glucopyranosyl-(1 → 6)-α-d-glucopyranosyl-(1 → 2)-β-d-fructofuranose. The ability of isomaltose to act as an acceptor in the Bacillus subtilis CECT 39 levansucrase-catalyzed transfructosylation reaction to efficiently produce theanderose in the presence of sucrose as a donor is described by using four different sucrose:isomaltose concentration ratios. The maximum theanderose concentration ranged from 122.4 to 130.4 g L
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I7253
Isomaltose, ~98% (TLC)
C12H22O11