EMAIL THIS PAGE TO A FRIEND
Scientific reports
Essential role of mitochondrial Stat3 in p38
PMID 29133922
Abstract
Stat3 is an oncogene, frequently associated with malignant transformation. A body of evidence implicates that phospho-Stat3