EMAIL THIS PAGE TO A FRIEND
The Journal of physiology
Regulation of TMEM16A/ANO1 and TMEM16F/ANO6 ion currents and phospholipid scrambling by Ca
PMID 29134661
Abstract
TMEM16 proteins can operate as Ca TMEM16/anoctamin (ANO) proteins form Ca