EMAIL THIS PAGE TO A FRIEND
Oncotarget
Molecular chaperones in the acquisition of cancer cell chemoresistance with mutated
PMID 29137250
Abstract
Utilizing the TCGA PANCAN12 dataset we discovered that cancer patients with mutations in