EMAIL THIS PAGE TO A FRIEND
Oncotarget
Inhibition of cell cycle progression by the hydroxytyrosol-cetuximab combination yields enhanced chemotherapeutic efficacy in colon cancer cells.
PMID 29137335
Abstract
Hydroxytyrosol (HT), a polyphenol of olive oil, downregulates epidermal growth factor (EGFR) expression and inhibits cell proliferation in colon cancer (CC) cells, with mechanisms similar to that activated by the EGFR inhibitor, cetuximab. Here, we studied whether HT treatment would enhance the cetuximab inhibitory effects on cell growth in CC cells. HT-cetuximab combination showed greater efficacy in reducing cell growth in HT-29 and WiDr cells at concentrations 10 times lower than when used as single agents. This reduction was clearly linked to cell cycle blockade, occurring at G
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
R6148 RNase A solution