EMAIL THIS PAGE TO A FRIEND
Oncotarget
Activated spleen tyrosine kinase promotes malignant progression of oral squamous cell carcinoma via mTOR/S6 signaling pathway in an ERK1/2-independent manner.
PMID 29137391
Abstract
Spleen tyrosine kinase (SYK), a non-receptor cytoplasmic tyrosine enzyme, is well known for its ability in certain pathways through immune receptors. Recently,
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
EHU081641 MISSION® esiRNA, esiRNA human SYK (esiRNA1)