EMAIL THIS PAGE TO A FRIEND
Nature communications
Quantitative analysis of protein-protein interactions and post-translational modifications in rare immune populations.
PMID 29142256
Abstract
In spite of recent advances in proteomics, quantitative analyses of protein-protein interactions (PPIs) or post-translational modifications (PTMs) in rare cell populations remain challenging. This is in particular true for analyses of rare immune and/or stem cell populations that are directly isolated from humans or animal models, and which are often characterized by multiple surface markers. To overcome these limitations, here we have developed proximity ligation imaging cytometry (PLIC), a protocol for proteomic analysis of rare cells. Specifically, by employing PLIC on medullary thymic epithelial cells (mTECs), which serve as a paradigm for a rare immune population, we demonstrate that PLIC overcomes the inherent limitations of conventional proteomic approaches and enables a high-resolution detection and quantification of PPIs and PTMs at a single cell level.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P1644 Percoll®, pH 8.5-9.5 (20 °C)