EMAIL THIS PAGE TO A FRIEND
Pharmacognosy magazine
High-performance Thin-layer Chromatographic-densitometric Quantification and Recovery of Bioactive Compounds for Identification of Elite Chemotypes of
PMID 29142436
Abstract
This study aimed to develop an easy, cheap, precise, and accurate high-performance thin-layer chromatographic (HPTLC) validated method for simultaneous quantification of bioactive alkaloids (colchicine and gloriosine) in The HPTLC chromatographic method was developed using mobile phase of chloroform: acetone: diethyl amine (5:4:1) at λ Five germplasms were collected from targeted region, and on morpho-anatomical inspection, no significant variation was observed among them. Quantification data reveal that content of colchicine ( The developed method is validated in terms of accuracy, recovery, and precision studies as per the ICH guidelines (2005) and can be adopted for the simultaneous quantification of colchicine and gloriosine in phytopharmaceuticals. In addition, this study is relevant to explore the chemotypic variability in metabolite content for commercial and medicinal purposes. An easy, cheap, precise, and accurate high performance thin layer chromatographic (HPTLC) validated method for simultaneous quantification of bioactive alkaloids (colchicine and gloriosine) in G. superba L.Five germplasms were collected from targeted region, and on morpho anatomical inspection, no significant variation was observed among themQuantification data reveal that content of colchicine (Rf: 0.72) and gloriosine (Rf: 0.61) varies from 0.035%-0.150% to 0.006%-0.032% (dry wt. basis)Two germplasms, namely NBG 27 and NBG 26, were found to be elite chemotype of both the markers.