EMAIL THIS PAGE TO A FRIEND
Stem cell research & therapy
Calcium-containing scaffolds induce bone regeneration by regulating mesenchymal stem cell differentiation and migration.
PMID 29145866
Abstract
Osteoinduction and subsequent bone formation rely on efficient mesenchymal stem cell (MSC) recruitment. It is also known that migration is induced by gradients of growth factors and cytokines. Degradation of Ca A circular calvarial bone defect (5 mm diameter) was created in the parietal bone of 35 Balb-C mice. We prepared and implanted a cell-free agarose/gelatin scaffold alone or in combination with different CaSO CaSO Specific CaSO