EMAIL THIS PAGE TO A FRIEND
Food chemistry
Effect of high-pressure processing on colour, phytochemical contents and antioxidant activities of purple waxy corn (Zea mays L. var. ceratina) kernels.
PMID 29146345
Abstract
High-pressure processing (HPP) at 250-700 MPa for 30-45 min affects the colour parameters, phytochemical contents and antioxidant activities of purple waxy corn kernels (p < 0.05). The higher pressure-level, the lower L