EMAIL THIS PAGE TO A FRIEND
Oncotarget
The
PMID 29152138
Abstract
First-line nilotinib in chronic myeloid leukemia is more effective than imatinib to achieve early and deep molecular responses, despite poor tolerability or failure observed in one-third of patients. The toxicity and efficacy of tyrosine kinase inhibitors might depend on the activity of transmembrane transporters. However, the impact of transporters genes polymorphisms in nilotinib setting is still debated. We investigated the possible correlation between single nucleotide polymorphisms of
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
CDS023093
Nilotinib
C28H22F3N7O