EMAIL THIS PAGE TO A FRIEND
Journal of neurology
T4, T3 and rT3 levels in serum and cerebrospinal fluid of patients with amyotrophic lateral sclerosis.
PMID 2915230
Abstract
Thyronine (T4), triiodothyronine (T3), and reverse-triiodothyronine (rT3) levels were evaluated in cerebrospinal fluid (CSF) and in serum of 12 patients with definite amyotrophic lateral sclerosis (ALS) by specific radioimmunoassays. Circulating microsomal and thyroglobulin antibodies were also evaluated. In all patients serum levels of T4, T3 and rT3 were within normal limits. In CSF, the rT3 levels were significantly elevated to 0.118 micrograms/l (mean), the T4 levels were not significantly elevated, and the T3 levels were below the detection limit of 0.03 micrograms/l. A correlation between the elevated rT3 levels in CSF and the severity or type of ALS could not be demonstrated by this study. The antithyroid antibodies (thyroglobulin antibodies, microsomal antibodies) showed normal titres and did not suggest disturbances of thyroid autoimmunity in the patients with ALS.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T5905
L-Thyronine
C15H15NO4