EMAIL THIS PAGE TO A FRIEND
Carbohydrate research
Synthesis of 3-aminopropyl β-(1 → 6)-d-glucotetraoside and its biotinylated derivative.
PMID 29156224
Abstract
3-Aminopropyl β-(1 → 6)-d-glucotetraoside has been synthesized from 3-benzyloxycarbonylaminopropanol and 6-O-acetyl-2,3,4-tri-O-benzoyl-d-glucopyranosyl trichloroacetimidate by successive attachment of one monosaccharide unit in total yield of 22%. Free aminopropyl glycoside was converted into a biotin derivative that can be used for controlled immobilization of the oligosaccharide on streptavidin-coated ELISA plates and for tracing carbohydrate binding molecules.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
447420
Orcinol, 97%
C7H8O2