EMAIL THIS PAGE TO A FRIEND
Oncotarget
miR-21 modulates the effect of EZH2 on the biological behavior of human lung cancer stem cells
PMID 29156731
Abstract
Non-small-cell lung cancer has a high mortality rate and poor prognosis. Therefore, novel therapeutic approaches are urgently needed to enhance patient survival rates. In this study, we investigated the effects of miR-21 and EZH2 on the biological behavior of human lung cancer stem cells
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
EHU073481 MISSION® esiRNA, esiRNA human EZH2 (esiRNA1)