EMAIL THIS PAGE TO A FRIEND
PeerJ
Ginsenoside Rk1 bioactivity: a systematic review.
PMID 29158964
Abstract
Ginsenoside Rk1 (G-Rk1) is a unique component created by processing the ginseng plant (mainly Sung Ginseng (SG)) at high temperatures. The aim of our study was to systematically review the pharmacological effects of G-Rk1. We utilized and manually searched eight databases to select